Learn More
Vector Laboratories Modification Buffer (10X) 5 x 1.5 mL
SDP
Catalog No. 502045754
Encompass
Modification Buffer is an amine-free, high buffering capacity solution which is optimized for the reaction of NHS esters and TFP esters with amino groups. Proteins, oligonucleotides, and other biomolecules are buffer exchanged into Modification Buffer prior to modification with S-HyNic and S-4FB to ensure efficient coupling of these linkers.
Catalog No. 50-204-5754 Supplier Vector Laboratories Supplier No. S4000005
Modification Buffer is an amine-free, high buffering capacity solution which is optimized for the reaction of NHS esters and TFP esters with amino groups. Proteins, oligonucleotides, and other biomolecules are buffer exchanged into Modification Buffer prior to modification with S-HyNic and S-4FB to ensure efficient coupling of these linkers.