Learn More
Sino Biological Peptide Substrates: Modified AKT Substrate
Catalog No. 502569866
Encompass_Preferred
The Modified AKT Substrate peptide sequence (modified-CKRPRAASFAE) is based on the N-terminus of GSK3.
Catalog No. 50-256-9866 Supplier Sino Biological Supplier No. A0558B
The Modified AKT Substrate peptide sequence (modified-CKRPRAASFAE) is based on the N-terminus of GSK3.