Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Device Modified Auramine O
SDP

Catalog No. 23769525 Shop All Scientific Device Laboratory Inc. Products
Change view
Click to view available options
Quantity:
250mL
1 x 4L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-769-525 250mL
23-769-524 1 x 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-769-525 Supplier Scientific Device Supplier No. 345250

missing translation for 'validMainframeMsg'

Fluorescent acid fast stain in two minutes

  • Permanganate step is omitted, only two step staining technique for acid fast organisms saves lot of time
  • For all non-acid fast organisms and debris, new counterstain quenches fluorescence
  • Yields cleaner field of view

Specifications

Quantity 250mL
Includes Stain and decolorizer or quencher
Format Bottle
For Use With (Application) Rapid and economical staining technique for acid fast organisms
Product Type Modified Auramine O Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.