Learn More
MOLTOX Molecular Toxicology Inc Modified Clostridial Broth, 100 mL/bottle, Store in Refrigerator
SDP
Catalog No. 50948769
Encompass_Preferred
Modified Clostridial Broth is used for the enumeration of Clostrida in food samples. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-769 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26577
Modified Clostridial Broth is used for the enumeration of Clostrida in food samples. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.