Learn More
Teknova Modified Columbia Agar with 5% Sheep's Blood. 100mm, 20 plates/sleeve. 4 pack.
SDP
Catalog No. 502387427
Encompass
1.20% Tryptone, 0.50% Peptone,0.35% Yeast Extract, 0.30% Beef Extract,0.10% Corn Starch, 0.50% Sodium Chloride,1.50% Agar, 5.00% Sheep's Blood
Catalog No. 50-238-7427 Supplier Teknova Supplier No. 2C41154PK
1.20% Tryptone, 0.50% Peptone,0.35% Yeast Extract, 0.30% Beef Extract,0.10% Corn Starch, 0.50% Sodium Chloride,1.50% Agar, 5.00% Sheep's Blood