Learn More
Electron Microscopy Sciences Modified Davidson's Fixative 1 LT
SDP
Catalog No. 5029228
Encompass_Preferred
Prepared from a combination of 14% Ethyl alcohol, Denatured, 37.5% Formalin 37-39%, Glacial Acetic Acid, and D I water.
Catalog No. 50-292-28 Supplier Electron Microscopy Sciences Supplier No. 6413350
Prepared from a combination of 14% Ethyl alcohol, Denatured, 37.5% Formalin 37-39%, Glacial Acetic Acid, and D I water.