Learn More
Bioworld MODIFIED DULBECCO S PBS PACKS
Catalog No. 50255873
Encompass_Preferred
Modified Dulbecco\'s PBS Packs are pouches of dry-blend powder that are each sufficient to make 500mL of physiological Na- and K-phosphate buffered saline (D-PBS) for ELISA and Western blot diluents and wash buffers. Dry-Blend Packs of m...
Catalog No. 50-255-873 Supplier Bioworld Supplier No. 404200121
Modified Dulbecco\'s PBS Packs are pouches of dry-blend powder that are each sufficient to make 500mL of physiological Na- and K-phosphate buffered saline (D-PBS) for ELISA and Western blot diluents and wash buffers. Dry-Blend Packs of m...