Learn More
Sigma Aldrich Fine Chemicals Biosciences Iscove’s Modified Dulbecco’s Medium, 500 mL
Catalog No. NC1623155
Encompass
Guilbert and Iscove demonstrated that precursor cells of erythrocytes and macrophages could be cultured in a totally defined serum free medium supplemented with albumin, transferrin, lecithin, and selenium. This medium is a modification of Dulbecco’s Modified Eagle’s Medium (DME) containing selenium, additional amino acids and vitamins, sodium pyruvate, HEPES buffer, and potassium nitrate instead of ferric nitrate.
Catalog No. NC1623155 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I3390500ML
Guilbert and Iscove demonstrated that precursor cells of erythrocytes and macrophages could be cultured in a totally defined serum free medium supplemented with albumin, transferrin, lecithin, and selenium. This medium is a modification of Dulbecco’s Modified Eagle’s Medium (DME) containing selenium, additional amino acids and vitamins, sodium pyruvate, HEPES buffer, and potassium nitrate instead of ferric nitrate.