Learn More
StatLab™ Modified EA-50 Cytology Stain
GSA_VASDP
Catalog No. STLSL9616
Change view
Click to view available options
Quantity:
4 x 1 gal.
4 x 16 oz. Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLSL9616 4 x 16 oz. Bottle
STLSL961 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL9616 Supplier StatLab™ Supplier No. SL9616
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
A popular substitute for EA-65.
  • Contains twice as much Eosin-Y than EA-50 and no Bismarck Brown
  • An alcohol-based counterstain used in the cytology laboratory for routine cytoplasmic staining
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 16 oz. Bottle
Product Type Cytology Stain