Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen Modified Harris Hematoxylin Stain

Catalog No. 23730607 Shop All Scigen Inc Products
Change view
Click to view available options
Quantity:
4/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-730-607 4/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-730-607 Supplier Scigen Supplier No. 7111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A regressive or progressive stain depending on your desired staining results, mercury-free, alcohol-free, no filtering required.

  • Modified Harris Hematoxylin produces excellent nuclear staining, nuclear chromatin is stained a crisp blue-purple with exceptional detail
  • Standard staining times range from 0.5 minutes to 3 minutes.
  • Staining intensity is controlled by increasing time increments by 30 seconds
  • Does not stain mucin
  • Does not form a metallic sheen or solids (precipitate), no filtering required Mercury free formulation
  • Certified biological stain
  • Manufactured using ISO 13485 and FDA quality systems

Specifications

Certifications/Compliance Traceable Certififed Biological Stain
Color Hematoxylin
Description Used in both regressive and progressive methods in histology and cytology preparations
For Use With (Application) Nuclear Staining
Includes Staining Protocol
Material Mercury-Free, Alcohol-Free
Packaging Light Resistant Bottle
Progressive/Regressive Both
Quantity 4/Cs.
Solution Type Aqueous
Storage Requirements Room Temperature, Dark Storage
Synonym Hematoxylin
Product Type Nuclear Stain
Show More Show Less

FOR IN VITRO LABORATORY USE POISON: NOT FOR DRUG, FOOD, OR HOUSEHOLD USE

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.