Learn More
Epredia™ Modified Harris Hematoxylin
Catalog No. 22050205 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-050-205 1 pt.
22-050-206 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-050-205 Supplier Epredia™ Supplier No. 72704
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use Epredia™ Modified Harris Hematoxylin, a regressive nuclear stain, for both histology and cytology.
  • Formulation contains no mercury
  • More stable than conventional Harris Hematoxylin
Specifications
Quantity 1 pt.
Product Type Nuclear Stain
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov