Learn More
MilliporeSigma Modified Hematoxylin Solution
Catalog No. M1051752500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1051752500 2.5 L
M1051750500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. M1051752500 Supplier MilliporeSigma Supplier No. 1051752500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hematoxylin solution modified according to Gill II for microscopy
Hematoxylin solution modified acc. to Gill II - for microscopy, is used for human-medical cell diagnosis and serves the purpose of the cytological and histological investigation of sample material of human origin. It is a ready-to-use staining solution that when used together with other in vitro diagnostic products from our portfolio makes cytological target structures evaluable for diagnostic purposes in human gynecological and clinico-cytological specimen materials, for example cervical smears. In hematoxylin-eosin staining, the most widely used histological staining technique, nuclear staining is achieved using ready-to-use staining solutions, such as Mayer's hemalum solution or Hematoxylin solution modified according to Gill II or Gill III. The dye is oxidized in a controlled manner, which renders the results robust and allows a brilliant differentiation of the stained structures. Counterstaining of i.e. for proteins, collagen, keratin, intercellular substances can be performed with alcoholic or aqueous Eosin Y solution 0.5%.These solutions do need to be filtrated prior use.
A 500 mL bottle is sufficient for 1500 to 2500 slides. It is registered as IVD and certified as CE product.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2.5 L
Product Type Hematoxylin Stain Solution