Learn More
Electron Microscopy Sciences Modified Hiraoka Staining Kit
SDP
Catalog No. 501931037
Encompass_Preferred
The modified Hiraoka Staining Kit stains up to 20 grids at a time. This modified kit features Hiraoka Staining Kit an autoclavable and microwavable the staining tray and staining tray insert. The modified plate holder as well as the parafilm well holder are made from Delrin, while the staining tray and staining tray insert are made from polypropylene. Includes Hiraoka Staining Tray (6), Plate Holder, Parafilm Well Holder, Space Insert (2), Grid Support Plate (2), Parafilm.
Catalog No. 50-193-1037 Supplier Electron Microscopy Sciences Supplier No. 7156000
The modified Hiraoka Staining Kit stains up to 20 grids at a time. This modified kit features Hiraoka Staining Kit an autoclavable and microwavable the staining tray and staining tray insert. The modified plate holder as well as the parafilm well holder are made from Delrin, while the staining tray and staining tray insert are made from polypropylene. Includes Hiraoka Staining Tray (6), Plate Holder, Parafilm Well Holder, Space Insert (2), Grid Support Plate (2), Parafilm.