Learn More
Electron Microscopy Sciences Modified Hiraoka Staining Tray, 46mm x 46mm x 11.5mm thick
SDP
Catalog No. 501931038
Encompass_Preferred
For use with the Modified Hiraoka Staining Kit. Made from polypropylene. Chemically resistant, autoclavable, and microwave safe.
Dimensions: 46mm x 46mm x 11.5mm thick
Catalog No. 50-193-1038 Supplier Electron Microscopy Sciences Supplier No. 7156010
For use with the Modified Hiraoka Staining Kit. Made from polypropylene. Chemically resistant, autoclavable, and microwave safe.
Dimensions: 46mm x 46mm x 11.5mm thick