Learn More
NEXPRING HEALTH Modified HTF Medium with Gentamicin - HEPES, 500 mL
Catalog No. NC1318490
Encompass
Modified HTF Medium is a synthetic, defined solution for procedures which include the retrieval, handling and transfer of human gametes and embryos. mHTF is composed of a HEPES and sodium bicarbonate buffering system, making it ideal for use in ambient atmosphere conditions. It requires protein supplementation before use.
Catalog No. NC1318490 Supplier NEXPRING HEALTH Supplier No. 90126500ML
Modified HTF Medium is a synthetic, defined solution for procedures which include the retrieval, handling and transfer of human gametes and embryos. mHTF is composed of a HEPES and sodium bicarbonate buffering system, making it ideal for use in ambient atmosphere conditions. It requires protein supplementation before use.