Learn More
NEXPRING HEALTH MODIFIED HTF MEDIUM 100/ML BTL
Catalog No. NC9425940
Encompass
Medium, Modified HTF; Irvine Scientific; Synthetic; w/Gentamicin-HEPES; Liquid; mHTF uses buffering system composed of 21mM HEPES, 4mM sodium bicarbonate combination; w/o CO2 incubation; Store 2 deg. to 8 deg.C; 100mL
Catalog No. NC9425940 Supplier NEXPRING HEALTH Supplier No. 90126100ML
Medium, Modified HTF; Irvine Scientific; Synthetic; w/Gentamicin-HEPES; Liquid; mHTF uses buffering system composed of 21mM HEPES, 4mM sodium bicarbonate combination; w/o CO2 incubation; Store 2 deg. to 8 deg.C; 100mL