Learn More
General Laboratory Products Modified Letheen Agar, 100 mL, 12/cs
Catalog No. NC1660165
Encompass_Preferred
Modified Letheen Agar - 100ml, Easy-pour narrow mouth glass bottle, 12 per case. Base for Microbiological Testing of Cosmetics and Bactericidal Activity of Quartenary Ammonium Compounds.
Catalog No. NC1660165 Supplier General Laboratory Products Supplier No. 125EZ7084100
Modified Letheen Agar - 100ml, Easy-pour narrow mouth glass bottle, 12 per case. Base for Microbiological Testing of Cosmetics and Bactericidal Activity of Quartenary Ammonium Compounds.