Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Modified Methyl Orange Indicator, 0.1% Methyl Orange-Xylene Cyanole, Reagents

Catalog No. 01334800
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-800 120 mL
01-334-797 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-800 Supplier Reagents Holdings Llc Supplier No. CM004100120A
Add to Cart
Add to Cart

Modified Methyl Orange Indicator, 0.1% Methyl Orange-Xylene Cyanole, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Modified Methyl Orange Indicator, 0.1% Methyl Orange-Xylene Cyanole
Boiling Point 100°C
CAS 7732-18-5, 2650-17-1
Synonym Methyl Orange-Xylene Cyanole
Percent Purity 0.1%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.001
Color Blue to Green
Quantity 120 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.