Learn More
StatLab Modified Trichrome Stain for Microsporidia
Description
- Trichrome stain can be used in conventional staining procedures
- Used in microbiology to visualize and differentiate various cellular structures, particularly in the context of protozoan infections of the gastrointestinal tract
- Can be used to detect and identify intestine protozoans from stool samples
- 250 mL bottle
Specifications
Specifications
| Includes | Trichrome Stain |
| Volume (Metric) | 250 mL |
| Compatible Cells | Microsporidia |
| Product Type | Trichrome Stain |