Learn More
StatLab Modified Trichrome Stain Set for Microsporidia
Description
- Used in microbiology to visualize and differentiate various cellular structures, particularly in the context of protozoan infections of the gastrointestinal tract
- Can be used to detect and identify intestine protozoans from stool samples
- Trichome stain and counterstain are used in microbiology for histological staining
- Trichome stain highlights collagen fibers and is used in the study of pathologies of the heart, liver, and kidney
- It increases the contrast of microscopic features in cells and tissues, making them easier to see when viewed through a microscope
- 250 mL bottle
Specifications
Specifications
| Includes | Trichrome Stain and Counterstain |
| Volume (Metric) | 250 mL |
| Compatible Cells | Microsporidia |
| Product Type | Trichrome Stain |