Learn More
Perkin Elmer US LLC Mg(NO3)2 Matrix Modifier for Graphite Furnace AA, 1% , 100 mL
Catalog No. 509042310
Encompass_Preferred
Bottle contains 100 mL of matrix modifier. Matrix modification decreases the volatility of the analyte element and prevents its loss during thermal pretreatment. This also allows application of higher pretreatment temperatures for better matrix removal. It also increases the volatility of matrix components and promotes their removal before atomization. PerkinElmer furnace autosamplers feature automatic matrix modification with the ability to apply one or more matrix modifiers sequentially or simultaneously. Refer to Technical Note “THGA Graphite Furnace AA – Automatic Matrix Modification for Improved Analytical Quality and Sample Throughput” for information on preparing the most common matrix modification solutions.
Catalog No. 50-904-2310 Supplier Perkin Elmer US LLC Supplier No. B0190634
Bottle contains 100 mL of matrix modifier. Matrix modification decreases the volatility of the analyte element and prevents its loss during thermal pretreatment. This also allows application of higher pretreatment temperatures for better matrix removal. It also increases the volatility of matrix components and promotes their removal before atomization. PerkinElmer furnace autosamplers feature automatic matrix modification with the ability to apply one or more matrix modifiers sequentially or simultaneously. Refer to Technical Note “THGA Graphite Furnace AA – Automatic Matrix Modification for Improved Analytical Quality and Sample Throughput” for information on preparing the most common matrix modification solutions.