Learn More
Medchemexpress LLC Modoflaner | 1331922-53-2 | Not found | 98.3% | C23H10F12IN3O2 | 10MG
SDP
Catalog No. 5000259837
Encompass_Preferred
Modoflaner is an isophenylamide insecticide research compound that may act through allosteric regulation of gamma-aminobutyric acid (GABA)-gated chloride channels. It is supplied as a white to off-white solid with reported high purity and is intended for research use only.
  • Isophenylamide insecticide acting via allosteric modulation of GABA-gated chloride channels.
  • Supplied as a white to off-white solid suitable for analytical and biological research.
  • High reported purity (≈98.3%) supports assay and screening applications.
  • Recommended storage conditions available for solid and solution to preserve stability.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-59837 Supplier Medchemexpress LLC Supplier No. HY13744510MG
Modoflaner is an isophenylamide insecticide research compound that may act through allosteric regulation of gamma-aminobutyric acid (GABA)-gated chloride channels. It is supplied as a white to off-white solid with reported high purity and is intended for research use only.
  • Isophenylamide insecticide acting via allosteric modulation of GABA-gated chloride channels.
  • Supplied as a white to off-white solid suitable for analytical and biological research.
  • High reported purity (≈98.3%) supports assay and screening applications.
  • Recommended storage conditions available for solid and solution to preserve stability.
  • Available in small research pack sizes for laboratory use.