Learn More
MSC MODUL-R GOGG W/VENT VISOR
Catalog No. 5000803581
Encompass
The Modul-R Safety Goggles offer extended coverage with an optional impact visor for additional protection. An open or closed venting system helps minimize fogging and can be flipped up in a stand-by position.
  • Low profile lens with extended field of vision
  • Strap pivots for added adjustability
  • Splash shield has an adjustable venting system to help minimize fog and splash
  • Anti-fog coating improves visibility under hot and humid conditions
  • Polycarbonate lens and frame material
  • Universal size
  • Offers 99.9% UV protection
  • Meets ANSI-ISEA Z87.1-2020 and CSA Z94.3 standards
  • Includes chin protector
  • Indirect venting system
Catalog No. 50-008-03581 Supplier MSC Supplier No. 72542897
The Modul-R Safety Goggles offer extended coverage with an optional impact visor for additional protection. An open or closed venting system helps minimize fogging and can be flipped up in a stand-by position.
  • Low profile lens with extended field of vision
  • Strap pivots for added adjustability
  • Splash shield has an adjustable venting system to help minimize fog and splash
  • Anti-fog coating improves visibility under hot and humid conditions
  • Polycarbonate lens and frame material
  • Universal size
  • Offers 99.9% UV protection
  • Meets ANSI-ISEA Z87.1-2020 and CSA Z94.3 standards
  • Includes chin protector
  • Indirect venting system