Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block with 40 Wells
SDP
Catalog No. 5032273
Encompass_Preferred
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 40 Well
Model: 12215-60
Catalog No. 50-322-73 Supplier Electron Microscopy Sciences Supplier No. 6313012
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 40 Well
Model: 12215-60