Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Modular Foley Rack, 2 Pocket Unit
SDP

Catalog No. 22000611
Change view
Click to view available options
Length (English):
21 in.
34 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-000-611 34 in.
22-000-610 21 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-611 Supplier Poltex Supplier No. FOLEYSTAK434
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Modular Foley Rack, 2 Pocket Unit is a modular Foley Catheter storage system.

  • 4 x 4 x 21 in. units combine with 4 x 4 x 34 in. units to create the perfect combination for all your storage needs.
  • Durable and cleanable PETG construction.
  • Each unit sold separately.

Specifications

Height (English) 4 in.
Length (English) 34 in.
Width (English) 4 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.