Learn More
Waters Corp Waters Modular Heat Block, For 1 mL Tubes, 96-Well Format
Catalog No. 502038292 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This modular heat block is designed to hold 1 mL tubes that come in strips of 8 within a 96-well format. It is specifically intended for use with the GlycoWorks RapiFluor-MS N-Glycan Kit workflow, utilizing the tubes provided in the Sample Collection Module.
  • Holds 1 mL tubes in strips of 8 in a 96-well format
  • Integrates into GlycoWorks RapiFluor-MS N-Glycan Kit workflow
  • Measures 5.8 in. long by 3.75 in. wide
  • Compatible with various dry bath heaters
Catalog No. 50-203-8292 Supplier Waters Corp Supplier No. 186007985
This modular heat block is designed to hold 1 mL tubes that come in strips of 8 within a 96-well format. It is specifically intended for use with the GlycoWorks RapiFluor-MS N-Glycan Kit workflow, utilizing the tubes provided in the Sample Collection Module.
  • Holds 1 mL tubes in strips of 8 in a 96-well format
  • Integrates into GlycoWorks RapiFluor-MS N-Glycan Kit workflow
  • Measures 5.8 in. long by 3.75 in. wide
  • Compatible with various dry bath heaters