Learn More
Electron Microscopy Sciences CoverWell™ Modular Hybridization System, 13 dia x 0.7 mm
SDP
Catalog No. 5030935
Encompass_Preferred
Flexible, press-to-seal gasket and coverslip enclosures which can be quickly assembled and disassembled for the specialized challenges of hybridizing and imaging thick and free- floating specimens.
All steps from permeabilizing to image analysis are made easy and trouble free with chambers you assembles from modular components.
For neural explants, plant, or animal specimens of any texture and thickness.
Build up to three chambers on one microscope slide. Ideally suited for overnight hybridizations and agitated washing steps. The screen is made from polyester and is 1.6 mm square.
Size: 13 dia x 0.7 mm
Catalog No. 50-309-35 Supplier Electron Microscopy Sciences Supplier No. 70334C
Flexible, press-to-seal gasket and coverslip enclosures which can be quickly assembled and disassembled for the specialized challenges of hybridizing and imaging thick and free- floating specimens.
All steps from permeabilizing to image analysis are made easy and trouble free with chambers you assembles from modular components.
For neural explants, plant, or animal specimens of any texture and thickness.
Build up to three chambers on one microscope slide. Ideally suited for overnight hybridizations and agitated washing steps. The screen is made from polyester and is 1.6 mm square.
Size: 13 dia x 0.7 mm