Learn More
Electron Microscopy Sciences CoverWell™ Modular Hybridization System, 13 dia x 0.7 mm
SDP
Supplier:  Electron Microscopy Sciences 70334D
Encompass_Preferred
Flexible, press-to-seal gasket and coverslip enclosures which can be quickly assembled and disassembled for the specialized challenges of hybridizing and imaging thick and free- floating specimens.
All steps from permeabilizing to image analysis are made easy and trouble free with chambers you assembles from modular components.
For neural explants, plant, or animal specimens of any texture and thickness.
Build up to three chambers on one microscope slide. Ideally suited for overnight hybridizations and agitated washing steps. The screen is made from polyester and is 1.6 mm square.
Size: 13 dia x 0.7 mm
Catalog No. 50-309-36