Learn More
Med Vet International Modular Osteotome Blades, 4mm, J1068MA
Catalog No. 5000585620
Encompass_Preferred
Modular Osteotome: a comfortable autoclavable teflon handle with metal end to take hammering Blade 1.1mm in thickness. This makes them a great choice for procedures like block sulcoplasty. The blades can be changed intra operatively for proper fit during the procedure. Supplied as a complete kit with 7 different blades or purchase handle and blades separately. Blade only 4mm
Catalog No. 50-005-85620 Supplier Med Vet International Supplier No. JOR1068MA
Modular Osteotome: a comfortable autoclavable teflon handle with metal end to take hammering Blade 1.1mm in thickness. This makes them a great choice for procedures like block sulcoplasty. The blades can be changed intra operatively for proper fit during the procedure. Supplied as a complete kit with 7 different blades or purchase handle and blades separately. Blade only 4mm