Learn More
Med Vet International Modular Osteotome Blades, 6mm, J1068MB
Catalog No. 5000585727
Encompass_Preferred
Modular Osteotome: a comfortable autoclavable teflon handle with metal end to take hammering Blade 1.1mm in thickness. This makes them a great choice for procedures like block sulcoplasty. The blades can be changed intra operatively for proper fit during the procedure. Supplied as a complete kit with 7 different blades or purchase handle and blades separately. Blade only 6mm
Catalog No. 50-005-85727 Supplier Med Vet International Supplier No. JOR1068MB
Modular Osteotome: a comfortable autoclavable teflon handle with metal end to take hammering Blade 1.1mm in thickness. This makes them a great choice for procedures like block sulcoplasty. The blades can be changed intra operatively for proper fit during the procedure. Supplied as a complete kit with 7 different blades or purchase handle and blades separately. Blade only 6mm