Learn More
Poltex Modular Serological Pipette Holder, 2 Pocket Unit
SDP
Catalog No. 22000614
Change view
Click to view available options
Height (English):
2.5 in.
3.5 in.
4.5 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
22-000-614 4.5 in.
22-000-612 2.5 in.
22-000-613 3.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-614 Supplier Poltex Supplier No. SRPIPSTAK245
Add to Cart
Add to Cart
The Modular Serological Pipette Holder, 2 Pocket Unit is a supplementary unit for Modular Serological Pipette Holder (two column version).
  • Choose from 2.5, 3.5, or 4.5 in. tall.
  • Attaches to themselves and/or Modular Serological Pipette Holder Starter Set (SRPIPSTAK2START).
  • See image for dimensions.
  • Comes with mounting hardware.
Specifications
Positions 2 Pockets
Height (English) 4.5 in.
Product Type Pipet Box Holder