Learn More
Cole-Parmer Modular stainless steel drying rack, 32 white pegs
Catalog No. NC2170842 Shop All Cole-Parmer Products
Encompass_Preferred
Removable "snap-in" pegs and add-on accessories let you turn these drying racks into customized work stations in just seconds!Corrosion-resistant 304 stainless steel (SS) racks and polypropylene pegs are easy to clean. Autoclavable.Integral catch drain directs all drainage into the drip trough—prevents accidental drainage to the back of the rack.Racks come with 6"L pegs, SS drain tube, and wall mounting bracket. Order add-on accessories, plugs, and additional pegs separately.Assorted peg colors include yellow, orange, pink, red, green, light blue, dark blue, lilac, and purple.
This item is not returnable. View return policy
Catalog No. NC2170842 Supplier Cole-Parmer Supplier No. 6703500
Removable "snap-in" pegs and add-on accessories let you turn these drying racks into customized work stations in just seconds!Corrosion-resistant 304 stainless steel (SS) racks and polypropylene pegs are easy to clean. Autoclavable.Integral catch drain directs all drainage into the drip trough—prevents accidental drainage to the back of the rack.Racks come with 6"L pegs, SS drain tube, and wall mounting bracket. Order add-on accessories, plugs, and additional pegs separately.Assorted peg colors include yellow, orange, pink, red, green, light blue, dark blue, lilac, and purple.