Learn More
Molecular Devices Molecular Devices Multi-Mode Validation Plate, Absorbance, Fluorescence, Luminescence
Catalog No. 5000014664
Encompass_Preferred
The SpectraTest Multi-Mode Validation Plate is an optical validation package that qualifies absorbance, fluorescence, luminescence, time-resolved fluorescence, and fluorescence polarization performance of compatible multi-mode microplate readers. SoftMax Pro software automates test execution, calculation, and reporting, and flags parameters that fall outside defined limits.
  • Validates absorbance, fluorescence, luminescence, time-resolved fluorescence, and fluorescence polarization.
  • Automates tests for linearity, precision, accuracy, and dynamic range.
  • Includes alignment checks, signal-to-noise and signal-to-background assessments, and filter checks.
  • Requires compatible analysis software version 7.0 or higher for automated protocols.
  • Recommended annual recertification to maintain traceability and performance.
Catalog No. 50-000-14664 Supplier Molecular Devices Supplier No. 02007200
Add to Cart
Add to Cart
Request a Quote
The SpectraTest Multi-Mode Validation Plate is an optical validation package that qualifies absorbance, fluorescence, luminescence, time-resolved fluorescence, and fluorescence polarization performance of compatible multi-mode microplate readers. SoftMax Pro software automates test execution, calculation, and reporting, and flags parameters that fall outside defined limits.
  • Validates absorbance, fluorescence, luminescence, time-resolved fluorescence, and fluorescence polarization.
  • Automates tests for linearity, precision, accuracy, and dynamic range.
  • Includes alignment checks, signal-to-noise and signal-to-background assessments, and filter checks.
  • Requires compatible analysis software version 7.0 or higher for automated protocols.
  • Recommended annual recertification to maintain traceability and performance.