Learn More
Alkali Scientific Moeller Decarboxylase Broth w/Ornithine 5mL Tube, 10/Pk
SDP
Catalog No. 5000663057
Encompass_Preferred
Moeller Decarboxylase Broth with Ornithine is a biochemical medium used to determine whether microorganisms produce the enzyme ornithine decarboxylase. The test is frequently used for differentiation of Gram negative enteric bacteria such as Enterobacter, Klebsiella, and Proteus species. Each 5 mL tube contains glucose, peptones, the amino acid ornithine, and pH indicators including bromocresol purple and cresol red. After inoculation, organisms ferment glucose and create an acidic environment that turns the medium yellow. If the organism possesses ornithine decarboxylase, the enzyme converts ornithine into the alkaline compound putrescine. This raises the pH and changes the medium back to purple. When the enzyme is absent, the medium remains yellow, indicating a negative reaction useful for biochemical identification.
Catalog No. 50-006-63057 Supplier Alkali Scientific Supplier No. MDMT02
Moeller Decarboxylase Broth with Ornithine is a biochemical medium used to determine whether microorganisms produce the enzyme ornithine decarboxylase. The test is frequently used for differentiation of Gram negative enteric bacteria such as Enterobacter, Klebsiella, and Proteus species. Each 5 mL tube contains glucose, peptones, the amino acid ornithine, and pH indicators including bromocresol purple and cresol red. After inoculation, organisms ferment glucose and create an acidic environment that turns the medium yellow. If the organism possesses ornithine decarboxylase, the enzyme converts ornithine into the alkaline compound putrescine. This raises the pH and changes the medium back to purple. When the enzyme is absent, the medium remains yellow, indicating a negative reaction useful for biochemical identification.