Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Moenomycin complex | 11015-37-5 | 99.9% | 10 MG
SDP

Catalog No. 5000438314
Encompass_Preferred

Moenomycin complex is a class of antibacterial agents and PBP inhibitors, effective against various bacterial PBPs. It functions by blocking the transglycosylase activity of class A penicillin-binding proteins (PBPs), inhibiting bacterial growth. This product is intended for research use in bacterial infections.

  • Appearance: Solid
  • Color: Off-white to light yellow
  • Solubility (in vitro): Ethanol : 25 mg/mL (requires ultrasonic and warming)
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months, -20°C for 1 month
  • Shipping: Room temperature in continental US; may vary elsewhere

Catalog No. 50-004-38314 Supplier Medchemexpress LLC Supplier No. HY12638710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Moenomycin complex is a class of antibacterial agents and PBP inhibitors, effective against various bacterial PBPs. It functions by blocking the transglycosylase activity of class A penicillin-binding proteins (PBPs), inhibiting bacterial growth. This product is intended for research use in bacterial infections.

  • Appearance: Solid
  • Color: Off-white to light yellow
  • Solubility (in vitro): Ethanol : 25 mg/mL (requires ultrasonic and warming)
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months, -20°C for 1 month
  • Shipping: Room temperature in continental US; may vary elsewhere

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.