Learn More
Medchemexpress LLC Moenomycin complex | 11015-37-5 | MFCD01742468 | 99.9% | 1583.6 g/mol | C69H107N4O35P | 5 MG
SDP
Catalog No. 5000264540
Encompass_Preferred
Moenomycin complex is a mixture of moenomycin antibiotics that potently inhibit bacterial transglycosylase enzymes, blocking the transglycosylase activity of class A penicillin-binding proteins (PBPs) and disrupting peptidoglycan biosynthesis. Supplied as a powder for research use, it is used in studies of bacterial cell wall synthesis, antibiotic mechanism of action, and in vitro antibacterial assays.
  • Potent transglycosylase inhibitor for mechanistic studies.
  • Inhibits class A penicillin-binding proteins, blocking cell wall biosynthesis.
  • High purity suitable for in vitro assays.
  • Supplied as a powder for convenient storage and handling.
  • Available in small pack sizes for scale-appropriate experiments.
  • Useful in antibacterial activity assays and biochemical characterization.
Catalog No. 50-002-64540 Supplier Medchemexpress LLC Supplier No. HY1263875MG
Add to Cart
Add to Cart
Moenomycin complex is a mixture of moenomycin antibiotics that potently inhibit bacterial transglycosylase enzymes, blocking the transglycosylase activity of class A penicillin-binding proteins (PBPs) and disrupting peptidoglycan biosynthesis. Supplied as a powder for research use, it is used in studies of bacterial cell wall synthesis, antibiotic mechanism of action, and in vitro antibacterial assays.
  • Potent transglycosylase inhibitor for mechanistic studies.
  • Inhibits class A penicillin-binding proteins, blocking cell wall biosynthesis.
  • High purity suitable for in vitro assays.
  • Supplied as a powder for convenient storage and handling.
  • Available in small pack sizes for scale-appropriate experiments.
  • Useful in antibacterial activity assays and biochemical characterization.