Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4478MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM562. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HT-29, CH3LC, or HUVEC cells. Uterus, placenta, tonsil (both B, T lymphocytes), skeletal muscle, thyroid, or kidney. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Moesin protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 78kDa.Chromosome: Xq11.1. Gene Symbol: MSN. Gene ID: 4478. Ensemble: ENSG00000147065. Uniprot: NP_002435.
Catalog No. 50-188-4945