Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4478MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: MSN/491. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HT-29, CH3LC, or HUVEC cells. Uterus, Placenta, Tonsil, Melanoma, Testicular Carcinoma. Cellular Staining: Cytoplasmic, Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human Moesin protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 78kDa.Chromosome: Xq11.1. Gene Symbol: MSN. Gene ID: 4478. Ensemble: ENSG00000147065. Uniprot: NP_002435.
Catalog No. 50-188-4944