Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Moexipril hydrochloride | 82586-52-5 | MFCD00940481 | 98.4% | 535.03 g/mol | C27H35ClN2O7 | 10 MG
SDP

Catalog No. 502082502
Encompass_Preferred

Moexipril hydrochloride is an angiotensin-converting enzyme inhibitor supplied as a white to off-white solid for laboratory research use. It is commonly used in preclinical pharmacology studies of blood pressure regulation and neuroprotection and is provided as a small sample suitable for assay development and in vitro testing.

  • CAS number: 82586-52-5.
  • Molecular weight: 535.03 g/mol.
  • Purity: 98.4% (typical, by HPLC).
  • Appearance: white to off-white solid.
  • Solubility: DMSO 250 mg/mL, water 100 mg/mL; ultrasonic recommended.
  • Storage: keep sealed and away from moisture; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Pack size: 10 mg vial suitable for research assays.

Catalog No. 50-208-2502 Supplier Medchemexpress LLC Supplier No. HYB0378A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Moexipril hydrochloride is an angiotensin-converting enzyme inhibitor supplied as a white to off-white solid for laboratory research use. It is commonly used in preclinical pharmacology studies of blood pressure regulation and neuroprotection and is provided as a small sample suitable for assay development and in vitro testing.

  • CAS number: 82586-52-5.
  • Molecular weight: 535.03 g/mol.
  • Purity: 98.4% (typical, by HPLC).
  • Appearance: white to off-white solid.
  • Solubility: DMSO 250 mg/mL, water 100 mg/mL; ultrasonic recommended.
  • Storage: keep sealed and away from moisture; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Pack size: 10 mg vial suitable for research assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.