Learn More
TARGETMOL CHEMICALS INC Moexipril hydrochloride 200MG
SDP
Catalog No. 502876893
Encompass_Preferred
Also available in 1 mL, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Moexipril hydrochloride (Moexipril HCl) is a potent orally active nonsulfhydrylangiotensin converting enzyme (ACE)inhibitor, used for the treatment of hypertension and congestive heart failure. Purity 89.27%
Catalog No. 50-287-6893 Supplier TARGETMOL CHEMICALS INC Supplier No. T6595200MG
Also available in 1 mL, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Moexipril hydrochloride (Moexipril HCl) is a potent orally active nonsulfhydrylangiotensin converting enzyme (ACE)inhibitor, used for the treatment of hypertension and congestive heart failure. Purity 89.27%