Learn More
Medchemexpress LLC Moexipril hydrochloride | 82586-52-5 | 98.4% | 100 MG
SDP
Catalog No. 5000407313
Encompass_Preferred
Moexipril hydrochloride is an orally active inhibitor of angiotensin-converting enzyme (ACE), and becomes effective by being hydrolyzed to moexiprila (hydrochloride). It exhibits antihypertensive and neuroprotective effects. This compound is for research use only and not sold to patients.
  • Inhibits angiotensin-converting enzyme (ACE)
  • Exhibits antihypertensive and neuroprotective effects
  • Significantly reduces the percentage of damaged neurons
  • Attenuates Fe2+/3+-induced neurotoxicity
  • Exhibits dose-dependent and antihypertensive effects in renal hypertensive rats, spontaneously hypertensive rats, and perinephritic hypertensive dogs
  • Significantly reduces the infarct area on the mouse brain surface
  • Significantly attenuates the cortical infarct volume in rats
Catalog No. 50-004-07313 Supplier Medchemexpress LLC Supplier No. HYB0378A100MG
Moexipril hydrochloride is an orally active inhibitor of angiotensin-converting enzyme (ACE), and becomes effective by being hydrolyzed to moexiprila (hydrochloride). It exhibits antihypertensive and neuroprotective effects. This compound is for research use only and not sold to patients.
  • Inhibits angiotensin-converting enzyme (ACE)
  • Exhibits antihypertensive and neuroprotective effects
  • Significantly reduces the percentage of damaged neurons
  • Attenuates Fe2+/3+-induced neurotoxicity
  • Exhibits dose-dependent and antihypertensive effects in renal hypertensive rats, spontaneously hypertensive rats, and perinephritic hypertensive dogs
  • Significantly reduces the infarct area on the mouse brain surface
  • Significantly attenuates the cortical infarct volume in rats