Learn More
Medchemexpress LLC Moexipril (hydrochloride) | 82586-52-5 | 98.4% | 25 MG
SDP
Catalog No. 5000451011
Encompass_Preferred
Moexipril hydrochloride is an orally active inhibitor of angiotensin-converting enzyme (ACE) that becomes effective after being hydrolyzed to moexiprilat (hydrochloride). This compound demonstrates significant antihypertensive and neuroprotective effects.
  • Orally active inhibitor of angiotensin-converting enzyme (ACE).
  • Becomes effective after hydrolysis to moexiprilat.
  • Exhibits antihypertensive and neuroprotective effects.
  • High potency against plasma ACE and purified ACE from rabbit lung.
  • Significantly reduces damaged neurons in a dose-dependent manner.
  • Attenuates Fe2+/3+-induced neurotoxicity.
  • Does not cause significant changes in apoptotic neurons.
  • Does not cross the blood-brain barrier.
  • Shows dose-dependent antihypertensive effects in various animal models.
  • Reduces infarct area on mouse brain surface.
  • Attenuates cortical infarct volume in rat brain.
Catalog No. 50-004-51011 Supplier Medchemexpress LLC Supplier No. 25MGHYB0378A
Moexipril hydrochloride is an orally active inhibitor of angiotensin-converting enzyme (ACE) that becomes effective after being hydrolyzed to moexiprilat (hydrochloride). This compound demonstrates significant antihypertensive and neuroprotective effects.
  • Orally active inhibitor of angiotensin-converting enzyme (ACE).
  • Becomes effective after hydrolysis to moexiprilat.
  • Exhibits antihypertensive and neuroprotective effects.
  • High potency against plasma ACE and purified ACE from rabbit lung.
  • Significantly reduces damaged neurons in a dose-dependent manner.
  • Attenuates Fe2+/3+-induced neurotoxicity.
  • Does not cause significant changes in apoptotic neurons.
  • Does not cross the blood-brain barrier.
  • Shows dose-dependent antihypertensive effects in various animal models.
  • Reduces infarct area on mouse brain surface.
  • Attenuates cortical infarct volume in rat brain.