Learn More
Medchemexpress LLC Moexipril hydrochloride | 82586-52-5 | 98.4% | 50 MG
SDP
Catalog No. 5000406092
Encompass_Preferred
Moexipril hydrochloride is an orally active inhibitor of angiotensin-converting enzyme (ACE) that becomes effective by being hydrolyzed to moexiprila (hydrochloride). It exhibits antihypertensive and neuroprotective effects and is for research use only.
  • Orally active ACE inhibitor
  • Exhibits antihypertensive effects
  • Exhibits neuroprotective effects
Catalog No. 50-004-06092 Supplier Medchemexpress LLC Supplier No. HYB0378A50MG
Moexipril hydrochloride is an orally active inhibitor of angiotensin-converting enzyme (ACE) that becomes effective by being hydrolyzed to moexiprila (hydrochloride). It exhibits antihypertensive and neuroprotective effects and is for research use only.
  • Orally active ACE inhibitor
  • Exhibits antihypertensive effects
  • Exhibits neuroprotective effects