Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moexipril hydrochloride, Tocris Bioscience™

Catalog No. 269150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-915-0 50 mg
26-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-915-0 Supplier R&D Systems Supplier No. 2691/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Angiotensin-converting enzyme (ACE) inhibitor

Angiotensin-converting enzyme (ACE) inhibitor that is hydrolyzed in the liver to the active metabolite moexiprilat (IC 50 values are 2.1 and 2700 nM for moexiprilat and moexipril respectively). Antihypertensive; decreases mean blood pressure in the spontaneous hypertensive rat (SHR). Also blocks degradation of bradykinin into inactive metabolites.

Chemical Identifiers

CAS 82586-52-5
Molecular Formula C27H35ClN2O7
Molecular Weight (g/mol) 535.03
MDL Number MFCD00940481
InChI Key JXRAXHBVZQZSIC-JKVLGAQCSA-N
Synonym moexipril hydrochloride, moexipril hcl, unii-q1umg3uh45, moexipril hydrochloride usan, q1umg3uh45, univasc tn, dsstox_cid_24267, dsstox_rid_80137, dsstox_gsid_44267
PubChem CID 54889
IUPAC Name hydrogen (3S)-2-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid chloride
SMILES [H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1CC2=CC(OC)=C(OC)C=C2C[C@H]1C(O)=O

Specifications

Quantity 50 mg
Formula Weight 535.03
Percent Purity >99%
Chemical Name or Material Moexipril hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.