Learn More
Cayman Chemical MoexIprIlhydrochlorIde 50mg
SDP
Catalog No. 502980993
Encompass_Preferred
A prodrug form of moexiprilat; inhibits ACE in a cell-free assay (IC50 = 2.7 µM); Inhibits PDE4 (IC50s = 38, 160, and 230 µM for PDE4B2, PDE4A5, and PDE4D5, respectively); reduces blood pressure in spontaneously hypertensive rats at 0.1-30 mg/kg per day; reduces infarct volume in a rat model of focal cerebral ischemia at 0.1 mg/kg
Catalog No. 50-298-0993 Supplier Cayman Chemical Supplier No. 2125550 MG
A prodrug form of moexiprilat; inhibits ACE in a cell-free assay (IC50 = 2.7 µM); Inhibits PDE4 (IC50s = 38, 160, and 230 µM for PDE4B2, PDE4A5, and PDE4D5, respectively); reduces blood pressure in spontaneously hypertensive rats at 0.1-30 mg/kg per day; reduces infarct volume in a rat model of focal cerebral ischemia at 0.1 mg/kg