Learn More
Medchemexpress LLC Mofegiline hydrochloride | 120635-25-8 | 99.6% | 233.69 | 1 ML
SDP
Catalog No. 5000392484
Encompass_Preferred
Mofegiline hydrochloride (MDL72974A) is an orally active and selective enzyme-activated irreversible inhibitor of MAO-B, demonstrating significant selectivity over MAO-A. It also functions as an inhibitor of semicarbazide-sensitive amine oxidase (SSAO). This compound is intended for research use only.
  • Selective enzyme-activated irreversible inhibitor of MAO-B.
  • Shows marked selectivity over MAO-A (IC50s of 3.6 nM for MAO-B and 680 nM for MAO-A).
  • Inhibits semicarbazide-sensitive amine oxidase (SSAO).
  • Inhibits [3H]dopamine uptake in rat striatum with an IC50 of 31.8 μM.
  • Inhibits SSAOs from various tissues including dog aorta, rat aorta, bovine aorta, and human umbilical artery.
Catalog No. 50-003-92484 Supplier Medchemexpress LLC Supplier No. HY16677A10MM/1ML
Mofegiline hydrochloride (MDL72974A) is an orally active and selective enzyme-activated irreversible inhibitor of MAO-B, demonstrating significant selectivity over MAO-A. It also functions as an inhibitor of semicarbazide-sensitive amine oxidase (SSAO). This compound is intended for research use only.
  • Selective enzyme-activated irreversible inhibitor of MAO-B.
  • Shows marked selectivity over MAO-A (IC50s of 3.6 nM for MAO-B and 680 nM for MAO-A).
  • Inhibits semicarbazide-sensitive amine oxidase (SSAO).
  • Inhibits [3H]dopamine uptake in rat striatum with an IC50 of 31.8 μM.
  • Inhibits SSAOs from various tissues including dog aorta, rat aorta, bovine aorta, and human umbilical artery.