Learn More
Medchemexpress LLC Mofegiline hydrochloride | 120635-25-8 | 99.6% | 233.69 | 100 MG
SDP
Catalog No. 5000392437
Encompass_Preferred
Mofegiline hydrochloride is an orally active and selective enzyme-activated irreversible inhibitor of MAO-B, showing significant selectivity over MAO-A. It is also an inhibitor of semicarbazide-sensitive amine oxidase (SSAO).
  • Orally active
  • Selective and irreversible inhibitor of MAO-B
  • Inhibits MAO-B with an IC50 of 3.6 nM
  • Less potent inhibition of MAO-A with an IC50 of 680 nM
  • Inhibits semicarbazide-sensitive amine oxidase (SSAO)
  • Demonstrates long half-life in daily dosing
  • Can block MPTP neurotoxicity in mice
Catalog No. 50-003-92437 Supplier Medchemexpress LLC Supplier No. HY16677A100MG
Mofegiline hydrochloride is an orally active and selective enzyme-activated irreversible inhibitor of MAO-B, showing significant selectivity over MAO-A. It is also an inhibitor of semicarbazide-sensitive amine oxidase (SSAO).
  • Orally active
  • Selective and irreversible inhibitor of MAO-B
  • Inhibits MAO-B with an IC50 of 3.6 nM
  • Less potent inhibition of MAO-A with an IC50 of 680 nM
  • Inhibits semicarbazide-sensitive amine oxidase (SSAO)
  • Demonstrates long half-life in daily dosing
  • Can block MPTP neurotoxicity in mice