Learn More
Cayman Chemical Mofegiline (hydrochloride), Purity: ≥95%, CAS: 120635-25-8, 50 mg
SDP
Catalog No. 502911026
Encompass_Preferred
An inhibitor of MAO-B (IC50 = 3.6 nM for the rat brain mitochondrial enzyme); selective for MAO-B over MAO-A (IC50 = 680 nM); inhibits VAP-1 (IC50 = 20 nM); reduces MPTP-induced decreases in striatal levels of dopamine, DOPAC, and HVA in mice at 1.25 mg/kg; inhibits LPS-induced increases in BALF levels of TNF-α in transgenic mice overexpressing VAP-1 at 5 mg/kg
Catalog No. 50-291-1026 Supplier Cayman Chemical Supplier No. 3501050 MG
An inhibitor of MAO-B (IC50 = 3.6 nM for the rat brain mitochondrial enzyme); selective for MAO-B over MAO-A (IC50 = 680 nM); inhibits VAP-1 (IC50 = 20 nM); reduces MPTP-induced decreases in striatal levels of dopamine, DOPAC, and HVA in mice at 1.25 mg/kg; inhibits LPS-induced increases in BALF levels of TNF-α in transgenic mice overexpressing VAP-1 at 5 mg/kg