Learn More
Apexbio Technology LLC MOG (35-55) 149635-73-4 1mg
SDP
Catalog No. 5000062224
Encompass_Preferred
MOG (35-55) (CAS 149635-73-4) is a truncated peptide derived from human myelin oligodendrocyte glycoprotein (MOG) MOG an immunoglobulin superfamily glycoprotein specifically expressed in the central nervous system serves as a target antigen in experimental autoimmune encephalomyelitis (EAE) an animal model resembling multiple sclerosis (MS) The MOG (35-55) peptide induces strong inflammatory responses via activation of autoreactive T cells and promotion of autoantibody production leading to extensive demyelination Due to its prominent encephalitogenic properties it is widely utilized in preclinical research for studying MS pathogenesis and therapeutic interventions
Catalog No. 50-000-62224 Supplier Apexbio Technology LLC Supplier No. A83061
MOG (35-55) (CAS 149635-73-4) is a truncated peptide derived from human myelin oligodendrocyte glycoprotein (MOG) MOG an immunoglobulin superfamily glycoprotein specifically expressed in the central nervous system serves as a target antigen in experimental autoimmune encephalomyelitis (EAE) an animal model resembling multiple sclerosis (MS) The MOG (35-55) peptide induces strong inflammatory responses via activation of autoreactive T cells and promotion of autoantibody production leading to extensive demyelination Due to its prominent encephalitogenic properties it is widely utilized in preclinical research for studying MS pathogenesis and therapeutic interventions