Learn More
Medchemexpress LLC MOG(35-55) amide, mouse, rat | 2022956-48-3 | 99.5% | 2580.97 | 25 MG
SDP
Catalog No. 5000397292
Encompass_Preferred
MOG(35-55) amide, mouse, rat is the terminal amidation form of the 35-55 fragment of the myelin oligodendrocyte glycoprotein immunogenic peptide. It can be utilized for experimental autoimmune encephalomyelitis (EAE) modeling.
  • Utilized for experimental autoimmune encephalomyelitis (EAE) modeling.
  • Expanded CD4+ T cells are largely specific for this peptide.
  • Induces a classic EAE model characterized by myelin sheath damage and flaccid paralysis in ABH mice.
  • Known encephalitis-inducing epitope in C57BL/6 mice.
Catalog No. 50-003-97292 Supplier Medchemexpress LLC Supplier No. 25MGHYP3719
MOG(35-55) amide, mouse, rat is the terminal amidation form of the 35-55 fragment of the myelin oligodendrocyte glycoprotein immunogenic peptide. It can be utilized for experimental autoimmune encephalomyelitis (EAE) modeling.
  • Utilized for experimental autoimmune encephalomyelitis (EAE) modeling.
  • Expanded CD4+ T cells are largely specific for this peptide.
  • Induces a classic EAE model characterized by myelin sheath damage and flaccid paralysis in ABH mice.
  • Known encephalitis-inducing epitope in C57BL/6 mice.